siRNA Details
| Gene Symbol: Gnpda1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011937.2 | NP_036067.2 | 2 | 138 | ||
| GenBank mRNA | AF088903.1 | - | 2 | 96 | ||
| AF160355.1 | - | 2 | 84 | |||
| AK080610.1 | - | 2 | 86 | |||
| AK084247.1 | - | 1 | 768 | |||
| BC014800.1 | - | 2 | 69 | 4220980 | ||
| BY092839.1 | - | 2 | 113 | |||
| CJ048539.1 | - | 2 | 139 | |||

Gnpda1
Genomic Map