siRNA Details
Gene Symbol: Ifi202b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008327.2 | NP_032353.2 | 5 | 1268 | ||
NM_011940.2 | NP_036070.2 | 5 | 1265 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ifi202b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008327.2 | NP_032353.2 | 5 | 1268 | ||
NM_011940.2 | NP_036070.2 | 5 | 1265 |