siRNA Details
| Gene Symbol: Ifi202b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008327.2 | NP_032353.2 | 2 | 251 | ||
| NM_011940.2 | NP_036070.2 | 2 | 248 | |||
| GenBank mRNA | BC018233.1 | - | 247 | 3990226 | ||
| CJ045697.1 | - | 2 | 202 | |||
| CJ056241.1 | - | 2 | 252 | |||
| M31418.1 | - | 320 | ||||

Ifi202b
Genomic Map