siRNA Details
| Gene Symbol: Ifi202b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008327.2 | NP_032353.2 | 2, 3 | 334 | ||
| NM_011940.2 | NP_036070.2 | 2, 3 | 331 | |||
| GenBank mRNA | AF140672.1 | - | 2, 3 | 204 | ||
| BC018233.1 | - | 330 | 3990226 | |||
| BC055888.1 | - | 1, 2 | 323 | 4945964 | ||
| CJ045697.1 | - | 2, 3 | 285 | |||
| CJ056241.1 | - | 2, 3 | 335 | |||
| M31418.1 | - | 403 | ||||

Ifi202b
Genomic Map