siRNA Details
Gene Symbol: Map3k12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163643.1 | NP_001157115.1 | 15 | 2870 | ||
NM_009582.4 | NP_033608.3 | 13 | 2694 | |||
GenBank mRNA | AK029882.1 | - | 13 | 2503 | ||
AK034374.1 | - | 15 | 2686 | |||
AK081623.1 | - | 8 | 1362 | |||
BC047158.1 | - | 12 | 2488 | 6494216 | ||
BC057572.1 | - | 15 | 2694 | 6401321 | ||
U14636.1 | - | 13 | 2526 |
