siRNA Details
| Gene Symbol: Nr5a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001159769.2 | NP_001153241.1 | 9 | 1660 | ||
| NM_030676.3 | NP_109601.1 | 9 | 1745 | |||
| GenBank mRNA | AB221629.1 | - | 9 | 1587 | ||
| AK054487.1 | - | 7 | 2165 | |||
| BC137845.1 | - | 9 | 1624 | 8860864 | ||
| DQ133942.1 | - | 9 | 1660 | |||
| M81385.1 | - | 9 | 1745 | |||

Nr5a2
Genomic Map