siRNA Details
| Gene Symbol: Slc27a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011978.2 | NP_036108.2 | 3 | 842 | ||
| GenBank mRNA | AF033031.1 | - | 3 | 723 | ||
| AJ223958.1 | - | 3 | 760 | |||
| AK144069.1 | - | 3 | 844 | |||
| AK144070.1 | - | 3 | 756 | |||
| AK165446.1 | - | 3 | 843 | |||
| BC013442.1 | - | 3 | 825 | 4236616 | ||
| BC022170.1 | - | 3 | 821 | 4159081 | ||
| BC024735.1 | - | 3 | 811 | 5123650 | ||

Slc27a2
Genomic Map