siRNA Details
Gene Symbol: Cyb5r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024195.2 | NP_077157.2 | 12 | 1263 | ||
GenBank mRNA | AF338818.1 | - | 11 | 1011 | ||
AK012855.1 | - | 11 | 902 | |||
AK034878.1 | - | 12 | 1207 | |||
AK078682.1 | - | 5 | 469 | |||
AK090159.1 | - | 12 | 1265 | |||
AK135261.1 | - | 11 | 914 | |||
AK157312.1 | - | 11 | 1267 | |||
AK167436.1 | - | 11 | 961 | |||
AK169937.1 | - | 11 | 1261 | |||
AK169995.1 | - | 11 | 1258 | |||
AK172252.1 | - | 11 | 1216 | |||
AK216438.1 | - | 2 | 163 | |||
AK217521.1 | - | 2 | 163 | |||
AY321368.1 | - | 10 | 1035 | |||
BC002170.1 | - | 5 | 463 | |||
BC025438.1 | - | 12 | 1274 | 5038563 |
