siRNA Details
| Gene Symbol: Snx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153680.2 | NP_710147.1 | 7 | 646 | ||
| GenBank mRNA | AK047265.1 | - | 6 | 740 | ||
| AK077650.1 | - | 7 | 648 | |||
| AK077780.1 | - | 7 | 648 | |||
| AK156299.1 | - | 7 | 697 | |||
| AK219724.1 | - | 4 | 291 | |||

Snx17
Genomic Map