siRNA Details
| Gene Symbol: Snx17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153680.2 | NP_710147.1 | 10 | 909 | ||
| GenBank mRNA | AK047265.1 | - | 9 | 1003 | ||
| AK077650.1 | - | 10 | 911 | |||
| AK077780.1 | - | 10 | 911 | |||
| AK156299.1 | - | 10 | 960 | |||
| BC023732.1 | - | 10 | 925 | 5352355 | ||
| BC023925.1 | - | 5 | 372 | |||
| BC026571.1 | - | 10 | 909 | 4986268 | ||
| BC029675.1 | - | 10 | 896 | |||

Snx17
Genomic Map