siRNA Details
| Gene Symbol: Ankrd13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172945.2 | NP_766533.2 | 13 | 1621 | ||
| GenBank mRNA | AK081149.1 | - | 299 | |||
| BC022693.1 | - | 3 | 333 | |||
| BC039786.1 | - | 3 | 806 | |||
| BC056928.1 | - | 8 | 850 | |||
| BC071189.1 | - | 7 | 707 | |||
| BC098207.1 | - | 6 | 660 | |||
| BC151158.1 | - | 13 | 1599 | 9088046 | ||
| BC172028.1 | - | 13 | 1600 | |||
| CD578820.1 | - | 7 | 717 | |||

Ankrd13b
Genomic Map