siRNA Details
| Gene Symbol: Ankrd13b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172945.2 | NP_766533.2 | 4, 5 | 554 | ||
| GenBank mRNA | BC151158.1 | - | 4, 5 | 532 | 9088046 | |
| BC172028.1 | - | 4, 5 | 532 | |||
| CA315262.1 | - | 3, 4 | 240 | |||
| CA329030.1 | - | 4, 5 | 542 | |||

Ankrd13b
Genomic Map