siRNA Details
| Gene Symbol: Rab11fip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175543.3 | NP_780752.1 | 7, 8 | 985 | ||
| GenBank mRNA | AK046734.1 | - | 7, 8 | 987 | ||
| AK147219.1 | - | 7, 8 | 1056 | |||
| AK147344.1 | - | 7, 8 | 1181 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rab11fip4| Gene Symbol: Rab11fip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175543.3 | NP_780752.1 | 7, 8 | 985 | ||
| GenBank mRNA | AK046734.1 | - | 7, 8 | 987 | ||
| AK147219.1 | - | 7, 8 | 1056 | |||
| AK147344.1 | - | 7, 8 | 1181 | |||