siRNA Details
| Gene Symbol: Eme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177752.4 | NP_808420.1 | 2 | 324 | ||
| GenBank mRNA | AK078516.1 | - | 2 | 325 | ||
| BC089459.1 | - | 2 | 299 | 30630337 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Eme1| Gene Symbol: Eme1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177752.4 | NP_808420.1 | 2 | 324 | ||
| GenBank mRNA | AK078516.1 | - | 2 | 325 | ||
| BC089459.1 | - | 2 | 299 | 30630337 | ||