siRNA Details
| Gene Symbol: E030025P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087087.1 | - | 1 | 481 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

E030025P04Rik| Gene Symbol: E030025P04Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK087087.1 | - | 1 | 481 | ||