siRNA Details
| Gene Symbol: Greb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252071.1 | NP_001239000.1 | 20 | 3445 | ||
| NM_015764.4 | NP_056579.2 | 22 | 3851 | |||
| GenBank mRNA | AB093248.1 | - | 5 | 828 | ||
| AF180470.1 | - | 21 | 3507 | |||
| AK147337.1 | - | 20 | 3379 | |||
| AK157808.1 | - | 20 | 3528 | |||
| BC160931.1 | - | 21 | 3377 | |||

Greb1
Genomic Map