siRNA Details
| Gene Symbol: Zfp759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172392.3 | NP_765980.3 | 4 | 1497 | ||
| GenBank mRNA | BC028265.1 | - | 4 | 1481 | 5365910 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp759| Gene Symbol: Zfp759 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172392.3 | NP_765980.3 | 4 | 1497 | ||
| GenBank mRNA | BC028265.1 | - | 4 | 1481 | 5365910 | |