siRNA Details
Gene Symbol: Fam107a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183187.3 | NP_899010.1 | 2 | 409 | ||
GenBank mRNA | AK044219.1 | - | 2 | 179 | ||
BC055107.1 | - | 2 | 409 | 6532624 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam107a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183187.3 | NP_899010.1 | 2 | 409 | ||
GenBank mRNA | AK044219.1 | - | 2 | 179 | ||
BC055107.1 | - | 2 | 409 | 6532624 |