siRNA Details
| Gene Symbol: Fam107a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183187.3 | NP_899010.1 | 2 | 444 | ||
| GenBank mRNA | AK044219.1 | - | 2 | 214 | ||
| AK165722.1 | - | 3 | 632 | |||
| BC055107.1 | - | 2 | 444 | 6532624 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam107a| Gene Symbol: Fam107a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183187.3 | NP_899010.1 | 2 | 444 | ||
| GenBank mRNA | AK044219.1 | - | 2 | 214 | ||
| AK165722.1 | - | 3 | 632 | |||
| BC055107.1 | - | 2 | 444 | 6532624 | ||