siRNA Details
Gene Symbol: Pclo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011995.4 | NP_036125.4 | 24 | 15321 | ||
GenBank mRNA | AB083477.1 | - | 23 | 15537 | ||
Y19185.2 | - | 25 | 15231 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Pclo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_011995.4 | NP_036125.4 | 24 | 15321 | ||
GenBank mRNA | AB083477.1 | - | 23 | 15537 | ||
Y19185.2 | - | 25 | 15231 |