siRNA Details
| Gene Symbol: Pclo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011995.4 | NP_036125.4 | 25 | 15403 | ||
| GenBank mRNA | AB083477.1 | - | 24 | 15619 | ||
| Y19185.2 | - | 26 | 15313 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pclo| Gene Symbol: Pclo | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011995.4 | NP_036125.4 | 25 | 15403 | ||
| GenBank mRNA | AB083477.1 | - | 24 | 15619 | ||
| Y19185.2 | - | 26 | 15313 | |||