siRNA Details
| Gene Symbol: Mroh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033365.2 | NP_001028537.1 | 2, 3 | 233 | ||
| GenBank mRNA | AK132992.1 | - | 552 | |||
| AK158548.1 | - | 2, 3 | 262 | |||
| AK158799.1 | - | 2, 3 | 234 | |||
| BC150895.1 | - | 1, 2 | 127 | 9087806 | ||
| BC172046.1 | - | 1, 2 | 127 | 9054332 | ||

Mroh5
Genomic Map