siRNA Details
| Gene Symbol: Xxylt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_198626.2 | NP_941028.2 | 3 | 843 | ||
| GenBank mRNA | AK154256.1 | - | 3 | 843 | ||
| AK166259.1 | - | 3 | 796 | |||
| AK195644.1 | - | 3 | 261 | |||
| BC031419.1 | - | 3 | 791 | 2615706 | ||

Xxylt1
Genomic Map