siRNA Details
| Gene Symbol: Robo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175549.4 | NP_780758.3 | 12 | 1766 | ||
| GenBank mRNA | AK035110.1 | - | 13 | 2652 | ||
| BC055333.1 | - | 13 | 2086 | 6409283 | ||
| DQ533875.1 | - | 14 | 2058 | |||
| DQ533876.1 | - | 12 | 1766 | |||

Robo2
Genomic Map