siRNA Details
| Gene Symbol: Med14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001048208.1 | NP_001041673.1 | 6 | 1024 | ||
| NM_012005.3 | NP_036135.3 | 6 | 1024 | |||
| GenBank mRNA | AB019028.1 | - | 6 | 1022 | ||
| AK084044.1 | - | 6 | 881 | |||
| AK163306.1 | - | 7 | 786 | |||
| BC065072.1 | - | 6 | 1124 | 6830820 | ||

Med14
Genomic Map