siRNA Details
| Gene Symbol: Arhgap28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172964.4 | NP_766552.3 | 12 | 1577 | ||
| GenBank mRNA | AK087523.1 | - | 12 | 1590 | ||
| AK133720.1 | - | 12 | 1416 | |||
| AK141901.1 | - | 12 | 1527 | |||
| AK162051.1 | - | 6 | 617 | |||
| BC066788.1 | - | 12 | 1567 | 30049271 | ||

Arhgap28
Genomic Map