siRNA Details
| Gene Symbol: Sap130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172965.2 | NP_766553.1 | 17 | 2559 | ||
| GenBank mRNA | AK032704.1 | - | 7 | 1224 | ||
| AK034401.1 | - | 17 | 2544 | |||
| AK053675.1 | - | 17 | 2560 | |||
| BC036975.1 | - | 7 | 1250 | |||
| BC063075.1 | - | 14 | 2086 | 5721433 | ||
| BC066030.1 | - | 17 | 2574 | |||

Sap130
Genomic Map