siRNA Details
| Gene Symbol: Zfp608 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175751.4 | NP_786927.2 | 6 | 4575 | ||
| GenBank mRNA | AK076178.1 | - | 2 | 1451 | ||
| AK077030.1 | - | 5 | 3302 | |||
| AK129321.1 | - | 1 | 252 | |||
| BC059268.1 | - | 5 | 4241 | |||
| BC092219.1 | - | 6 | 4308 | 6830664 | ||
| BC151073.1 | - | 5 | 4094 | 9087986 | ||
| BC151075.1 | - | 5 | 4094 | 9087988 | ||

Zfp608
Genomic Map