siRNA Details
| Gene Symbol: Sh2d1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012009.4 | NP_036139.3 | 270 | |||
| GenBank mRNA | AB196832.1 | - | 218 | |||
| AB196833.1 | - | 218 | ||||
| AB196834.1 | - | 218 | ||||
| AB196835.1 | - | 218 | ||||
| AF020263.1 | - | 218 | ||||
| BC116698.1 | - | 270 | 40105516 | |||
| BC117508.1 | - | 270 | 40105515 | |||
| BC145540.1 | - | 276 | 9054103 | |||
| DQ131180.1 | - | 218 | ||||
| DQ132507.1 | - | 218 | ||||
| DQ132508.1 | - | 218 | ||||
| DQ132509.1 | - | 218 | ||||
| DQ132510.1 | - | 218 | ||||

Sh2d1b1
Genomic Map