siRNA Details
| Gene Symbol: Mgat4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290801.1 | NP_001277730.1 | 14, 15 | 1820 | ||
| NM_173870.3 | NP_776295.1 | 15, 16 | 1899 | |||
| GenBank mRNA | AB053217.1 | - | 14, 15 | 1575 | ||
| AK052278.1 | - | 12, 13 | 1396 | |||
| AK155724.1 | - | 15, 16 | 1901 | |||
| AK163327.1 | - | 14, 15 | 1652 | |||

Mgat4a
Genomic Map