siRNA Details
| Gene Symbol: Cntrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012018.2 | NP_036148.2 | 3 | 402 | ||
| GenBank mRNA | AK020148.1 | - | 3 | 386 | ||
| AK031338.1 | - | 3 | 385 | |||
| AK041715.1 | - | 3 | 403 | |||
| BC038386.1 | - | 3 | 380 | |||
| BC158010.1 | - | 2 | 275 | 9007318 | ||

Cntrl
Genomic Map