siRNA Details
| Gene Symbol: Cntrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012018.2 | NP_036148.2 | 43 | 7133 | ||
| GenBank mRNA | AK084186.1 | - | 3 | 930 | ||
| AK131139.1 | - | 21 | 3487 | |||
| BC079536.1 | - | 31 | 5158 | |||
| BC158010.1 | - | 42 | 7006 | 9007318 | ||

Cntrl
Genomic Map