siRNA Details
| Gene Symbol: Ccdc32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_199310.2 | NP_955014.2 | 2, 3 | 337 | ||
| GenBank mRNA | AK040670.1 | - | 2, 3 | 307 | ||
| AK132604.1 | - | 2, 3 | 329 | |||
| AK133813.1 | - | 2, 3 | 311 | |||
| AK147152.1 | - | 2, 3 | 313 | |||
| BC023926.1 | - | 2, 3 | 331 | 5324154 | ||
| BY086759.1 | - | 2, 3 | 338 | |||
| BY262121.1 | - | 2, 3 | 343 | |||

Ccdc32
Genomic Map