siRNA Details
Gene Symbol: Epb4.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001128606.1 | NP_001122078.1 | 13 | 2174 | ||
NM_001128607.1 | NP_001122079.1 | 13 | 2084 | |||
NM_183428.3 | NP_906273.3 | 16 | 3598 | |||
GenBank mRNA | AB370255.1 | - | 1 | 48 | ||
AB370261.1 | - | 13 | 2016 | |||
AK133437.1 | - | 3 | 369 | |||
AK147266.1 | - | 8 | 1127 | |||
AK147644.1 | - | 8 | 1127 | |||
AK220462.1 | - | 13 | 2081 | |||
BC018450.1 | - | 1 | 70 | |||
BC031511.1 | - | 1 | 70 | |||
BC043337.1 | - | 2 | 221 | |||
BC068138.1 | - | 13 | 2044 | |||
BC079875.1 | - | 13 | 2003 | 6844082 | ||
CB526050.1 | - | 6 | 541 | |||
L00919.1 | - | 17 | 3673 | |||
LK021332.1 | - | 12 | 1392 | |||
LK021333.1 | - | 12 | 1392 |
