siRNA Details
| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031393.2 | NP_113570.2 | 5, 6 | 611 | ||
| GenBank mRNA | AK078868.1 | - | 5, 6 | 600 | ||
| AK131162.1 | - | 5, 6 | 568 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Sytl1| Gene Symbol: Sytl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031393.2 | NP_113570.2 | 5, 6 | 611 | ||
| GenBank mRNA | AK078868.1 | - | 5, 6 | 600 | ||
| AK131162.1 | - | 5, 6 | 568 | |||