siRNA Details
| Gene Symbol: Cnpy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175651.4 | NP_783582.1 | 3 | 637 | ||
| GenBank mRNA | AK035828.1 | - | 3 | 638 | ||
| AK139155.1 | - | 2 | 1323 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cnpy1| Gene Symbol: Cnpy1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175651.4 | NP_783582.1 | 3 | 637 | ||
| GenBank mRNA | AK035828.1 | - | 3 | 638 | ||
| AK139155.1 | - | 2 | 1323 | |||