siRNA Details
| Gene Symbol: Cul1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012042.3 | NP_036172.1 | 9 | 1291 | ||
| GenBank mRNA | AF083216.1 | - | 8 | 1015 | ||
| AF136441.1 | - | 8 | 1146 | |||
| AF176910.1 | - | 9 | 1180 | |||
| AK147004.1 | - | 9 | 1273 | |||
| AK164274.1 | - | 9 | 1258 | |||
| AK165978.1 | - | 9 | 1232 | |||
| AK209075.1 | - | 2 | 154 | |||
| BC004836.1 | - | 8 | 1100 | |||
| BC029260.1 | - | 9 | 1291 | 2812400 | ||

Cul1
Genomic Map