siRNA Details
Gene Symbol: Gm15800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181421.4 | NP_852086.3 | 46 | 7377 | ||
GenBank mRNA | AK044167.1 | - | 13 | 1886 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm15800 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181421.4 | NP_852086.3 | 46 | 7377 | ||
GenBank mRNA | AK044167.1 | - | 13 | 1886 |