siRNA Details
| Gene Symbol: Pla2g2f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_012045.4 | NP_036175.2 | 1 | 167 | ||
| GenBank mRNA | AF166099.2 | - | 1 | 166 | ||
| AK029254.1 | - | 1 | 157 | |||
| BC125567.1 | - | 1 | 68 | 40130082 | ||
| BY146964.1 | - | 1 | 168 | |||

Pla2g2f
Genomic Map