siRNA Details
| Gene Symbol: Aak1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001040106.2 | NP_001035195.1 | 14 | 2609 | ||
| NM_177762.6 | NP_808430.2 | 13 | 2366 | |||
| GenBank mRNA | AK129272.1 | - | 4 | 827 | ||
| AK147363.1 | - | 14 | 2610 | |||
| BC043125.1 | - | 3 | 415 | |||
| BC141176.1 | - | 14 | 2365 | 9055788 | ||

Aak1
Genomic Map