siRNA Details
| Gene Symbol: Vat1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173016.3 | NP_766604.2 | 3 | 580 | ||
| GenBank mRNA | AK122527.1 | - | 3 | 580 | ||
| BC056927.1 | - | 3 | 590 | 5719211 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vat1l| Gene Symbol: Vat1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173016.3 | NP_766604.2 | 3 | 580 | ||
| GenBank mRNA | AK122527.1 | - | 3 | 580 | ||
| BC056927.1 | - | 3 | 590 | 5719211 | ||