siRNA Details
| Gene Symbol: Ccdc153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081369.2 | NP_001074838.2 | 6 | 465 | ||
| GenBank mRNA | BC038167.1 | - | 4 | 516 | ||
| BG072259.2 | - | 2 | 618 | |||
| CX240611.1 | - | 6 | 442 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc153| Gene Symbol: Ccdc153 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081369.2 | NP_001074838.2 | 6 | 465 | ||
| GenBank mRNA | BC038167.1 | - | 4 | 516 | ||
| BG072259.2 | - | 2 | 618 | |||
| CX240611.1 | - | 6 | 442 | |||