siRNA Details
| Gene Symbol: Myo9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173018.2 | NP_766606.2 | 25 | 5164 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo9a| Gene Symbol: Myo9a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173018.2 | NP_766606.2 | 25 | 5164 | ||