siRNA Details
| Gene Symbol: Klhl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177771.5 | NP_808439.2 | 3 | 379 | ||
| GenBank mRNA | AK044774.1 | - | 3 | 490 | ||
| BC065167.1 | - | 3 | 275 | |||
| BY195998.1 | - | 3 | 300 | |||
| BY248410.1 | - | 3 | 299 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Klhl18| Gene Symbol: Klhl18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177771.5 | NP_808439.2 | 3 | 379 | ||
| GenBank mRNA | AK044774.1 | - | 3 | 490 | ||
| BC065167.1 | - | 3 | 275 | |||
| BY195998.1 | - | 3 | 300 | |||
| BY248410.1 | - | 3 | 299 | |||