siRNA Details
Gene Symbol: Taf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290729.1 | NP_001277658.1 | 29, 30 | 4609 | ||
GenBank mRNA | AB299229.1 | - | 29, 30 | 4536 | ||
AK046668.1 | - | 8, 9 | 1098 | |||
BC047418.1 | - | 19, 20 | 2722 | |||
BC094568.1 | - | 11, 12 | 1893 |
