siRNA Details
Gene Symbol: B9d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013717.2 | NP_038745.1 | 7 | 587 | ||
GenBank mRNA | AB030483.1 | - | 7 | 587 | ||
AK003038.1 | - | 7 | 582 | |||
AK010355.1 | - | 7 | 616 | |||
AK131701.1 | - | 7 | 578 | |||
BC008113.1 | - | 7 | 555 | 3483517 |
