siRNA Details
Gene Symbol: Zbtb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173026.2 | NP_766614.2 | 4 | 1554 | ||
GenBank mRNA | AK033794.1 | - | 1 | 235 | ||
AK048360.1 | - | 1 | 1835 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zbtb11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173026.2 | NP_766614.2 | 4 | 1554 | ||
GenBank mRNA | AK033794.1 | - | 1 | 235 | ||
AK048360.1 | - | 1 | 1835 |