siRNA Details
| Gene Symbol: Zbtb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173026.2 | NP_766614.2 | 5 | 1945 | ||
| GenBank mRNA | AK033794.1 | - | 2 | 626 | ||
| AK048360.1 | - | 2 | 2226 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zbtb11| Gene Symbol: Zbtb11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173026.2 | NP_766614.2 | 5 | 1945 | ||
| GenBank mRNA | AK033794.1 | - | 2 | 626 | ||
| AK048360.1 | - | 2 | 2226 | |||