siRNA Details
Gene Symbol: 4933408B17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177773.4 | NP_808441.2 | 1 | 1409 | ||
GenBank mRNA | AK030167.1 | - | 1 | 1409 | ||
BC172130.1 | - | 1 | 1283 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4933408B17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177773.4 | NP_808441.2 | 1 | 1409 | ||
GenBank mRNA | AK030167.1 | - | 1 | 1409 | ||
BC172130.1 | - | 1 | 1283 |