siRNA Details
| Gene Symbol: 4933408B17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177773.4 | NP_808441.2 | 3 | 1659 | ||
| GenBank mRNA | AK030167.1 | - | 3 | 1659 | ||
| BC172130.1 | - | 3 | 1533 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

4933408B17Rik| Gene Symbol: 4933408B17Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_177773.4 | NP_808441.2 | 3 | 1659 | ||
| GenBank mRNA | AK030167.1 | - | 3 | 1659 | ||
| BC172130.1 | - | 3 | 1533 | |||